Sodium hydroxide 98%, pellets, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

13407-0010 13407-0100 134070025 134070250
TS13407-0010EA 113 USD
TS13407-0010 TS13407-0100 TS134070025 TS134070250
Sodium hydroxide 98%, pellets, extra pure
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance White Pellets
Titration with HCl ≥97.5 %
Chloride (Cl) ≤50 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤40 ppm
Sodium carbonate (Na2CO3) ≤1.0 %
Aluminium (Al) ≤10 ppm
Calcium (Ca) ≤10 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤1 ppm
Nickel (Ni) ≤5 ppm
Potassium (K) ≤250 ppm
Silver (Ag) ≤0.5 ppm
Silicon dioxide (SiO2) ≤5 ppm
Nitrogen compounds (as N) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR