Hydrogen chloride 5->6 N in 2-propanol, pure

Supplier: Thermo Scientific Chemicals
Danger

13370-0010 133700025
TS13370-0010EA 216 USD
TS13370-0010 TS133700025
Hydrogen chloride 5->6 N in 2-propanol, pure
Hydrogen chloride
Formula: HCl
MW: 36.46
Density: 0.909 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2920
ADR: 8 I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Water ≤1.0 %
Titration with NaOH 5 to 6 N
NMR (31P-NMR) Consistent with structure
Elemental Analysis 4.9 to 5.3 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR