Hydrogen chloride 5->6 N in 2-propanol, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
13370-0010
133700025
TS13370-0010EA
216
USD
TS13370-0010
TS133700025
Hydrogen chloride 5->6 N in 2-propanol, pure
Hydrogen chloride
Formula:
HCl MW: 36.46 Density: 0.909 g/cm³ Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 2920 ADR: 8 I |
Specification Test Results
Appearance | Clear colorless to light yellow Solution |
Water | ≤1.0 % |
Titration with NaOH | 5 to 6 N |
NMR (31P-NMR) | Consistent with structure |
Elemental Analysis | 4.9 to 5.3 % N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...