Sulfanilamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Aminobenzenesulfonamide, 4-Aminobenzenesulphonamide

13285-0025 13285-5000 13285-1000
TS13285-0025EA 629 USD
TS13285-0025 TS13285-5000 TS13285-1000
Sulfanilamide 98%
Sulphanilamide
Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: 401
Melting Pt: 163…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS Number: 63-74-1
EINECS: 200-563-4
Merck Index: 13,09010
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bb9ab6bdf23e9d846b24d4f57047dea1d5071&vid=f2f1c5e47a127cd33d3b4a2a31b1057c375456eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 163 to 167 °C
Loss on drying ≤0.5 % (1 g, 105 °C)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR