Sebacic acid 98%

Supplier: Thermo Scientific Chemicals

13263-0010 13263-0025 13263-0100 13263-1000
TS13263-0010EA 128 USD
TS13263-0010 TS13263-0025 TS13263-0100 TS13263-1000
Sebacic acid 98%
Sebacic acid
Formula: C₁₀H₁₈O₄
Boiling Pt: 294.4 °C (100 mmHg)
Melting Pt: 131…134 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
REACH: 01-2119519212-52
Merck Index: 12,08558

Order Now

Specification Test Results

Appearance White to off-white Powder or granules
Infrared spectrum Conforms
Water ≤0.3 %
GC ≥97.5 %
Ash ≤0.06 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR