D-(-)-Ribose 99+%

Supplier: Thermo Scientific Chemicals

13236-0050 13236-0250 13236-1000 13236-5000
TS13236-0050EA 29 USD
TS13236-0050 TS13236-0250 TS13236-1000 TS13236-5000
D-(-)-Ribose 99+%
D(-)-Ribose
Formula: C₅H₁₀O₅
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371

Order Now

Specification Test Results

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (40°C, 5 h, 20 Torr)
Sulfated ash ≤0.1 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -19.2° to -20.8° (20°C, 589 nm) (c=4, H2O)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR