Propyl-3,4,5-trihydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

13158-0025 13158-1000 13158-5000
TS13158-0025EA 801 USD
TS13158-0025 TS13158-1000 TS13158-5000
Propyl-3,4,5-trihydroxybenzoate 98%
Propyl 3,4,5-trihydroxybenzoate
Formula: C₁₀H₁₂O₅
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002196
CAS Number: 121-79-9
EINECS: 204-498-2
Merck Index: 12,08044

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 150 °C
Loss on drying ≤0.5 % (110 °C, 4 hrs)
Sulfated ash ≤0.1 %
Titration with NaOH 97.5 to 102.5 % (On dry substance)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR