4-Phenylbutyric acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Phenylbutanoic acid

13038-0250 13038-1000 13038-5000
TS13038-0250EA 67 USD
TS13038-0250 TS13038-1000 TS13038-5000
4-Phenylbutyric acid 99%
4-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
Boiling Pt: 165 °C (10 mmHg)
Melting Pt: 49…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8

Order Now

Specification Test Results

Appearance White to light yellow to beige Powder or crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 49 to 53 °C
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR