2-Indolinone 97%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone

12967-0050 12967-0250 12967-1000
TS12967-0050EA 53 USD
TS12967-0050 TS12967-0250 TS12967-1000
2-Indolinone 97%, pure
2-Indolinone
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 313
Melting Pt: 123…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5

Order Now

Specification Test Results

Appearance Beige to yellow to orange to pink-brown Powder
Infrared spectrum Conforms
Melting point 123 to 128 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR