1,8-Octanediol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,8-Dihydroxy octane

12938-0250 12938-1000
TS12938-0250EA 41 USD
TS12938-0250 TS12938-1000
1,8-Octanediol 98%
1,8-Octanediol
Formula: C₈H₁₈O₂
Boiling Pt: 278.8 °C (760 mmHg)
Melting Pt: 60…62 °C
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002989
CAS Number: 629-41-4
EINECS: 211-090-8

Order Now

Specification Test Results

Appearance Colorless to white Powder or crystals
Infrared spectrum Conforms
Melting point 58 to 63 °C
Water ≤0.5 % (K.F.)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR