3-Methylthiophene 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12786-0500
12786-2500
TS12786-0500EA
67
USD
TS12786-0500
TS12786-2500
3-Methylthiophene 99+%
3-Methylthiophene
Formula:
C₅H₆S Boiling Pt: 115…117 °C (760 mmHg) Melting Pt: –69 °C Density: 1.016 g/cm³ Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005470 CAS Number: 616-44-4 EINECS: 210-482-6 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5180 to 1.5210 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...