1-Methylpiperidine 99%

Supplier: Thermo Scientific Chemicals
Danger

12748-0010 12748-0100 12748-2500
TS12748-0010EA 238 USD
TS12748-0010 TS12748-0100 TS12748-2500
1-Methylpiperidine 99%
1-Methylpiperidine
Formula: C₆H₁₃N
Boiling Pt: 105…108 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.82 g/cm³
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00006491
CAS Number: 626-67-5
EINECS: 210-959-9
UN: 2399
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4368 to 1.4388 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR