4-Methyl-1-pentene 97%

Supplier: Thermo Scientific Chemicals
Danger

12742-0250
TS12742-0250EA 80 USD
TS12742-0250
4-Methyl-1-pentene 97%
4-Methyl-1-pentene
Formula: C₆H₁₂
Boiling Pt: 54 °C (1013 hPa)
Melting Pt: –155 °C
Density: 0.66 g/cm³
Flash Pt: –31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008949
CAS Number: 691-37-2
EINECS: 211-720-1
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3810 to 1.3830 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR