(±)-3-Methylcyclohexanone 97%

Supplier: Thermo Scientific Chemicals
Warning

12663-0050 12663-1000 12663-5000
TS12663-0050EA 25 USD
TS12663-0050 TS12663-1000 TS12663-5000
(±)-3-Methylcyclohexanone 97%
(±)-3-Methylcyclohexanone
Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 169
Melting Pt: -73
Density: 0.915 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00001639
CAS Number: 591-24-2
EINECS: 209-710-7
UN: 2297
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR