Mercaptoacetic acid (thioglycolic acid) 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercaptoacetic acid

12543-2500 12543-0010 12543-0025 12543-0100
TS12543-2500EA 88 USD
TS12543-2500 TS12543-0010 TS12543-0025 TS12543-0100
Mercaptoacetic acid (thioglycolic acid) 99+%, extra pure
Mercaptoacetic acid (Thioglycolic acid)
Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 96
Melting Pt: -16.5
Density: 0.99 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 201-176-3
UN: 3463
ADR: 8 II
REACH: 01-2119486971-24
Merck Index: 13,09410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45179825145ec44f2e058197a0ee54845ae7c750&vid=51e43f5b69b880b196570b3065f9b4d8db7336d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3870 (20°C, 589 nm)
Water ≤0.1 %
GC ≥99.0 %
Color scale ≤10 APHA
UV 1 cm cell vs water
Residue after evaporation ≤0.001 %
Acidity ≤0.0003 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR