DL-Menthol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol

12539-0050 12539-1000 12539-5000
TS12539-0050EA 38 USD
TS12539-0050 TS12539-1000 TS12539-5000
DL-Menthol 99%
DL-Menthol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Melting Pt: 32…36 °C
Density: 0.89 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00001484
CAS Number: 89-78-1
EINECS: 201-939-0

Order Now

Specification Test Results

Appearance Colorless to white Crystalline solid
Infrared spectrum Conforms
Melting point 32 to 36 °C
GC ≤0.1 % 2-Propanol
Specific optical rotation -2° to +2° (20 °C, 589 nm) (c=10, EtOH)
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Acidity (CH3COOH) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR