D-(-)-Mannitol 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
12534-0010
12534-0025
12534-0050
12534-1000
12534-5000
TS12534-0010EA
79
USD
TS12534-0010
TS12534-0025
TS12534-0050
TS12534-1000
TS12534-5000
D-(-)-Mannitol 98+%
D(-)-Mannitol
Formula:
HOH₂C(CH(OH))₄CH₂OH MW: 182.17 g/mol Melting Pt: 165…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064287 CAS Number: 69-65-8 EINECS: 200-711-8 Merck Index: 13,05769 |
|
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 165 to 170 °C |
Loss on drying | ≤0.3 % (105 °C, 4 hrs) |
HPLC | ≥98.0 % (ELSD) |
Specific optical rotation | +23° to +25° (20 °C, 589 nm) (Ph.Eur.-directives) |
Nickel (Ni) | ≤1 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...