DL-Malic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

12525-0050 12525-0010 12525-2500 12525-5000
TS12525-0050EA 191 USD
TS12525-0050 TS12525-0010 TS12525-2500 TS12525-5000
DL-Malic acid 99+%
DL-Malic acid
Formula: HOOCCH(OH)CH₂COOH
Melting Pt: 128…132 °C
Flash Pt: 203 °C
Storage Temperature: Ambient
MDL Number: MFCD00064212
CAS Number: 617-48-1
EINECS: 210-514-9
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 128 to 132 °C
Sulfated ash ≤0.1 %
Specific optical rotation -0.1° to +0.1° (25 °C, 589 nm) (C=8.5%,H2O)
Titration with NaOH ≥99.0 %
Lead (Pb) ≤2 ppm
Impurity ≤0.05 % Maleic acid
Insoluble matter ≤0.1 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR