Iron(III) chloride hexahydrate 99+%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Iron trichloride hexahydrate, Iron trichloride hexahydrate, Iron (III) chloride 6-hydrate, Iron trichloride 6-hydrate, Ferric chloride 6-hydrate, Ferric chloride hexahydrate, Ferric chloride hexahydrate
12503-0250
AC12503-0010
AC12503-0025
AC12503-0050
TS12503-0250EA
1014
USD
TS12503-0250
TS12503-0010
TS12503-0025
TS12503-0050
Iron(III) chloride hexahydrate 99+%, extra pure
Iron (III) chloride hexahydrate
Formula:
FeCl₃·6H₂O Boiling Pt: 280…285 °C Melting Pt: 37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149712 CAS Number: 10025-77-1 EINECS: 231-729-4 UN: 3260 ADR: 8,III Merck Index: 13,04048 |
|
Specification Test Results
Appearance | Yellow to brown Granules or lumps |
Titration Iodometric | ≥99.0 % |
Sulfate (SO4) | ≤1500 ppm |
Ferrous iron (Fe 2+) | ≤0.9 % |
Acidity (HCl) | ≤0.8 % |
Insoluble matter | ≤0.05 % (10 g in 50 ml H2O:HCl (49:1)) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...