Disodium hydrogen phosphate dihydrate 98+% for analysis

Supplier: Thermo Scientific Chemicals

AC12499-0010 AC12499-0050 AC12499-2500
TS12499-0010EA 94 USD
TS12499-0010 TS12499-0050 TS12499-2500
Disodium hydrogen phosphate dihydrate 98+% for analysis
di-Sodium hydrogen phosphate dihydrate
Formula: Na₂HPO₄·2H₂O
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7

Order Now

Specification Test Results

Appearance White Powder or crystals
Loss on drying 19.5 % to 21.0 % (1 g, 130°C)
Solubility (5 % in water) Clear colorless
pH 8.4 to 9.6 (1% in water)
Titration excess HCl 98.0 to 102.0 %
Sulfates ≤0.1 %
Chlorides ≤0.04 %
Arsenic (As) ≤0.0003 %
Cadmium (Cd) ≤0.005 %
Iron (Fe) ≤0.004 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.3 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR