Sulfuric acid ∼96% in water for analysis

Supplier: Thermo Scientific Chemicals
Danger

12464-0010 124640011 124640025 124640026 124640100 124645000 124645001
TS12464-0010EA 68 USD
TS12464-0010 TS124640011 TS124640025 TS124640026 TS124640100 TS124645000 TS124645001
Sulfuric acid ∼96% in water for analysis
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,08974

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Residue after ignition ≤0.0005 %
Titration with NaOH 95 to 97 %
Chloride (Cl) ≤0.5 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.5 ppm
Heavy metals (as Pb) ≤2 ppm
KMnO4-reducing substance ≤0.0008 % (as SO2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR