Chromone-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Oxo-4H-1-benzopyran-2-carboxylic acid, Chromocarb, 4-Oxo-4H-chromene-2-carboxylic acid

12388-0050 12388-0250
TS12388-0050EA 37 USD
TS12388-0050 TS12388-0250
Chromone-2-carboxylic acid 97%
Chromone-2-carboxylic Acid
Formula: C₁₀H₆O₄
Melting Pt: 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006838
CAS Number: 4940-39-0
EINECS: 225-583-0

Order Now

Specification Test Results

Appearance White to beige to grey-brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR