Tris(perfluorobutyl)amine , Perfluoro-compound FC-43™

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

12377-0100
TS12377-0100EA 102 USD
TS12377-0100 TS12377-1000 TS12377-2500
Tris(perfluorobutyl)amine , Perfluoro-compound FC-43™
Tris(perfluorobutyl)amine
Formula: C₁₂F₂₇N
MW: 671.1
Boiling Pt: 155
Melting Pt: -55
Density: 1.88 g/cm³
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Boiling point 173 to 181 °C
Density (25 °C) 1.865 to 1.895 g/ml
Additional info dielectric strength: > 35 kV
Viscosity 2.3 to 3.3 cSt (at 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR