2-Pentanone 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl Propyl Ketone

12350-2500 123500010 123500025 123500100
TS12350-2500EA 52 USD
TS12350-2500 TS123500010 TS123500025 TS123500100
2-Pentanone 99%, pure
2-Pentanone
Formula: C₅H₁₀O
Boiling Pt: 100…103 °C (760 mmHg)
Melting Pt: –78 °C
Density: 0.809 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009400
CAS Number: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3,II
Merck Index: 12,06193

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3910 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR