2',3'-O-Isopropylideneadenosine 98%
Supplier: Thermo Scientific Chemicals
12272-0050
12272-0250
TS12272-0050EA
66
USD
TS12272-0050
TS12272-0250
2',3'-O-Isopropylideneadenosine 98%
2',3'-O-Isopropylideneadenosine
Formula:
C₁₃H₁₇N₅O₄ Melting Pt: 221…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005756 CAS Number: 362-75-4 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
UV spectrophotometry | molar absorptivity 15400 +/-4%, on dry substance |
Ratio of Absorbances | ≤0.01 (A290/A260) |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...