2',3'-O-Isopropylideneadenosine 98%

Supplier: Thermo Scientific Chemicals

12272-0050 12272-0250
TS12272-0050EA 66 USD
TS12272-0050 TS12272-0250
2',3'-O-Isopropylideneadenosine 98%
2',3'-O-Isopropylideneadenosine
Formula: C₁₃H₁₇N₅O₄
Melting Pt: 221…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00005756
CAS Number: 362-75-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV spectrophotometry molar absorptivity 15400 +/-4%, on dry substance
Ratio of Absorbances ≤0.01 (A290/A260)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR