5-Iodo-2'-deoxyuridine (Idoxuridine) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine

12235-0010 12235-0050 12235-0250
TS12235-0010EA 82 USD
TS12235-0010 TS12235-0050 TS12235-0250
5-Iodo-2'-deoxyuridine (Idoxuridine) 99%
5-Iodo-2'-deoxyuridine (Idoxuridine)
Formula: C₉H₁₁IN₂O₅
MW: 354.10 g/mol
Melting Pt: 155…180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00134656
CAS Number: 54-42-2
EINECS: 200-207-8

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 60°C) (vacuum)
Specific optical rotation +28° to +32° (20°C, 589 nm) (c=1, 1 N NaOH) (on dried substance)
Titration with NaOH ≥98.5 % (On dried substance)
Oxalate (C2O4) (limit about 0.05%) Passes test
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤3 ppm
Lead (Pb) ≤2 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR