Iodoacetamide 99%

Supplier: Thermo Scientific Chemicals
Warning

12227-1000 12227-0050 12227-0250 12227-5000
TS12227-1000EA 674 USD
TS12227-1000 TS12227-0050 TS12227-0250 TS12227-5000
Iodoacetamide 99%
Iodoacetamide
Formula: ICH₂CONH₂
Boiling Pt: 168…170 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.871 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-572-7
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4110 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 0.869 to 0.873
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR