Indole 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1H-Indole

12215-0025 12215-0100 12215-0250 12215-1000 12215-5000
TS12215-0025EA 844 USD
TS12215-0025 TS12215-0100 TS12215-0250 TS12215-1000 TS12215-5000
Indole 99+%
Indole
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253…254 °C (760 mmHg)
Melting Pt: 51…53 °C
Density: 1.22 g/cm³
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,04985

Order Now

Specification Test Results

Appearance White to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 53 °C
GC ≥99.0 %
UV at 195 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR