Indene 90% stabilised, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Indene
12214-0010
12214-0050
12214-2500
TS12214-0010EA
180
USD
TS12214-0010
TS12214-0050
TS12214-2500
Indene 90% stabilised, Technical Grade
Indene
Formula:
C₉H₈ Boiling Pt: 181 °C (760 mmHg) Melting Pt: 5…3 °C Density: 0.99 g/cm³ Flash Pt: 56 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003777 CAS Number: 95-13-6 EINECS: 202-393-6 UN: 3295 ADR: 3,III Merck Index: 13,04964 |
Specification Test Results
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5670 to 1.5750 (20°C, 589 nm) |
GC | ≥88 % |
Stabilizer | 5 to 10 ppm tert-Butylcatechol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...