Dopamine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
12200-0100
12200-0250
12200-1000
TS12200-0100EA
81
USD
TS12200-0100
TS12200-0250
TS12200-1000
Dopamine hydrochloride 99%
Dopamine hydrochloride
Formula:
C₈H₁₂ClNO₂ Melting Pt: 241…243 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012898 CAS Number: 62-31-7 EINECS: 200-527-8 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (105°C, 2 hrs) |
Sulfated ash | ≤0.1 % |
Titration with HClO4 | ≥98.5 % (On dried substance) |
Heavy metals (as Pb) | ≤20 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...