1-(2-Hydroxyethyl)pyrrolidine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Pyrrolidineethanol, N-(2-Hydroxyethyl)pyrrolidine

12120-0250 12120-1000 12120-5000
TS12120-0250EA 57 USD
TS12120-0250 TS12120-1000 TS12120-5000
1-(2-Hydroxyethyl)pyrrolidine 95%
1-(2-Hydroxyethyl)pyrrolidine
Formula: C₆H₁₃NO
Boiling Pt: 79…81 °C (13 mmHg)
Density: 0.98 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003181
CAS Number: 2955-88-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4713 to 1.4743 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR