1-(2-Hydroxyethyl)pyrrolidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Pyrrolidineethanol, N-(2-Hydroxyethyl)pyrrolidine
12120-0250
12120-1000
12120-5000
TS12120-0250EA
57
USD
TS12120-0250
TS12120-1000
TS12120-5000
1-(2-Hydroxyethyl)pyrrolidine 95%
1-(2-Hydroxyethyl)pyrrolidine
Formula:
C₆H₁₃NO Boiling Pt: 79…81 °C (13 mmHg) Density: 0.98 g/cm³ Flash Pt: 56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003181 CAS Number: 2955-88-6 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear light yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4713 to 1.4743 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...