Hydroquinone 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,4-Dihydroxybenzene, Quinol
12091-0020
12091-0100
12091-5000
TS12091-0020EA
144
USD
TS12091-0020
TS12091-0100
TS12091-5000
Hydroquinone 99%
Hydroquinone
Formula:
C₆H₄(OH)₂ MW: 110.11 g/mol Boiling Pt: 285 Melting Pt: 172 Flash Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002339 CAS Number: 123-31-9 EINECS: 204-617-8 UN: 3077 ADR: 9 III Merck Index: 13,04833 |
|
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals or needles |
Infrared spectrum | Conforms |
Melting point | 170 to 175 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...