Hydroquinone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Dihydroxybenzene, Quinol

12091-0020 12091-0100 12091-5000
TS12091-0020EA 144 USD
TS12091-0020 TS12091-0100 TS12091-5000
Hydroquinone 99%
Hydroquinone
Formula: C₆H₄(OH)₂
MW: 110.11 g/mol
Boiling Pt: 285
Melting Pt: 172
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002339
CAS Number: 123-31-9
EINECS: 204-617-8
UN: 3077
ADR: 9 III
Merck Index: 13,04833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203cfb319003b2b4489cbe3ab15876fc37f27cb8&vid=6d76260c677491e292c03637f8a08e9194282d48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 170 to 175 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR