Sorbic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid

12053-0010 12053-1000 12053-2500 12053-5000
TS12053-0010EA 138 USD
TS12053-0010 TS12053-1000 TS12053-2500 TS12053-5000
Sorbic acid 99%
Sorbic acid
Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
Boiling Pt: 228
Melting Pt: 132…135 °C
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
Merck Index: 13,08793
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 132 to 135 °C
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.2 %
Titration with NaOH 98.5 to 101.5 % (On anhydrous substance)
Arsenic (As) ≤3 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR