n-Hexadecane 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexadecane, Cetane

12046-0025 12046-0100 12046-5000 12046-1000
TS12046-0025EA 667 USD
TS12046-0025 TS12046-0100 TS12046-5000 TS12046-1000
n-Hexadecane 99%, pure
n-Hexadecane
Formula: H₃C(CH₂)₁₄CH₃
Boiling Pt: 287 °C (760 mmHg)
Melting Pt: 18 °C
Density: 0.773 g/cm³
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid or solid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR