Glycoluril 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrahydroimidazo[4,5-d]imidazole-2,5(1H,3H)-dione, Acetylenediurea

12013-0250 12013-1000 12013-5000
TS12013-0250EA 38 USD
TS12013-0250 TS12013-1000 TS12013-5000
Glycoluril 97%
Glycoluril
Formula: C₄H₆N₄O₂
MW: 142.12 g/mol
Boiling Pt: 862 °C (760 mmHg)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005539
CAS Number: 496-46-8

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Assay ≥96.0 % (N)
NMR consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR