Fumaric acid 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11975-0010
11975-0025
11975-0100
11975-1000
TS11975-0010EA
55
USD
TS11975-0010
TS11975-0025
TS11975-0100
TS11975-1000
Fumaric acid 99+%
Fumaric acid
Formula:
C₄H₄O₄ Melting Pt: 295…300 °C (subl.) Density: 1.62 g/cm³ Flash Pt: 230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002700 CAS Number: 110-17-8 EINECS: 203-743-0 Merck Index: 13,04308 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 295 to 300 °C |
Water | ≤0.25 % (K.F.) |
HPLC | ≥99.0 % |
Titration with NaOH | 99.0 to 101.0 % (On anhydrous substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...