Ethylene carbonate 99+%

Supplier: Thermo Scientific Chemicals
Warning

11841-0010 11841-0051 11841-2500
TS11841-0010EA 60 USD
TS11841-0010 TS11841-0051 TS11841-2500
Ethylene carbonate 99+%
Ethylene carbonate
Formula: C₃H₄O₃
Boiling Pt: 246.7 °C (760 mmHg)
Melting Pt: 34…37 °C
Density: 1.321 g/cm³
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005382
CAS Number: 96-49-1
EINECS: 202-510-0

Order Now

Specification Test Results

Appearance White to yellow Low melting crystalline solid
Infrared spectrum Conforms
Melting point 34 to 37 °C
GC ≥99.0 %
Specific optical rotation +66.3° to +66.8° (25 °C, 589 nm) (c=26, H2O)
Chloride (Cl) ≤50 ppm
SO4 and SO3 (as SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Titrable acid ≤0.0008 meq/g
Insoluble matter ≤0.005 %
Invert sugar ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR