(±)-Ethyl-2-bromovalerate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-bromovalerate

118160250 118161000
TS118160250EA 30 USD
TS118160250 TS118161000
(±)-Ethyl-2-bromovalerate 99%
(±)-Ethyl 2-bromovalerate
Formula: C₇H₁₃BrO₂
MW: 209.09 g/mol
Density: 1.22 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00000159
CAS Number: 615-83-8
EINECS: 210-450-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4476 to 1.4496 (20 °C, 589 nm)
GC ≥98.5 %
Iodate (IO3) ≤5 ppm
Nitrite (NO2) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤30 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤20 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR