2-Ethylaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11805-0010
11805-0050
11805-2500
TS11805-0010EA
198
USD
TS11805-0010
TS11805-0050
TS11805-2500
2-Ethylaniline 98%
2-Ethylaniline
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 210 Melting Pt: -46 Density: 0.98 g/cm³ Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007751 CAS Number: 578-54-1 EINECS: 209-424-2 UN: 2273 ADR: 6.1 III |
Specification Test Results
Appearance | Clear yellow to red-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5590 to 1.5610 (20°C, 589 nm) |
GC | ≥97.5 % |
Chloride (Cl) | ≤0.001 % |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤0.002 % |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤5 ppm |
Magnesium (Mg) | ≤0.002 % |
Potassium (K) | ≤0.005 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
KMnO4-reducing substance | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...