2,2'-Bipyridyl 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
11750-0100
11750-0250
11750-1000
11750-5000
TS11750-0100EA
51
USD
TS11750-0100
TS11750-0250
TS11750-1000
TS11750-5000
2,2'-Bipyridyl 99+%
2,2'-Bipyridyl
Formula:
C₁₀H₈N₂ Boiling Pt: 272…273 °C (760 mmHg) Melting Pt: 69…72 °C Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006212 CAS Number: 366-18-7 EINECS: 206-674-4 UN: 2811 ADR: 6.1,III Merck Index: 1303382 |
|
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 69 to 72 °C |
GC | ≥99.0 % |
Suitability | (for Fe determination) Passes test |
Barium (Ba) | ≤0.001 % |
Calcium (Ca) | ≤0.01 % |
Iron (Fe) | ≤5 ppm |
Manganese (Mn) | ≤0.001 % |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.005 % |
Strontium (Sr) | ≤0.005 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...