2,5-Diphenyloxazole 99% for scintillation

Supplier: Thermo Scientific Chemicals

11738-5000 11738-0025 11738-0250 11738-1000
TS11738-5000EA 384 USD
TS11738-5000 TS11738-0025 TS11738-0250 TS11738-1000
2,5-Diphenyloxazole 99% for scintillation
2,5-Diphenyloxazole
Formula: C₁₅H₁₁NO
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 70…72 °C
Density: 1.06 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005306
CAS Number: 92-71-7
EINECS: 202-181-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 72 °C
Additional info background: < 25 cpm
HPLC ≥98.5 %
UV transmittance ≥85 % (40% in toluene)
Color of solution ≤50 APHA (40% in toluene)
Carbonyl compounds ≤0.002 % (acetone)
Titrable acid or base ≤0.0001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR