Docusate sodium 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Dioctyl sulfosuccinate sodium salt, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulfonate, Sodium dioctyl sulfosuccinate, Dioctyl sulfosuccinate, sodium salt, bis(2-Ethylhexyl) sodium sulfosuccinate

11710-1000 11710-0010 11710-0025
TS11710-1000EA 57 USD
TS11710-1000 TS11710-0010 TS11710-0025
Docusate sodium 96%
Docusate sodium
Formula: C₂₀H₃₇NaO₇S
MW: 444.57 g/mol
Melting Pt: 155
Storage Temperature: Ambient
MDL Number: MFCD00012455
CAS Number: 577-11-7
EINECS: 209-406-4
Merck Index: 12,03460

Order Now

Specification Test Results

Appearance White Waxy solid
Infrared spectrum Conforms
Assay ≥95 %
Water ≤2 %
Color of alcohol solution ≤100 APHA (50 % solids in 1:1 alcohol/water)
Insoluble matter ≤300 ppm (50 % in toluene)
Iodine value ≤0.25 (on solids basis)
Acid number ≤2.5 (on solids basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR