Docusate sodium 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Dioctyl sulfosuccinate sodium salt, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulfonate, Sodium dioctyl sulfosuccinate, Dioctyl sulfosuccinate, sodium salt, bis(2-Ethylhexyl) sodium sulfosuccinate
11710-1000
11710-0010
11710-0025
TS11710-1000EA
57
USD
TS11710-1000
TS11710-0010
TS11710-0025
Docusate sodium 96%
Docusate sodium
Formula:
C₂₀H₃₇NaO₇S MW: 444.57 g/mol Melting Pt: 155 Storage Temperature: Ambient |
MDL Number:
MFCD00012455 CAS Number: 577-11-7 EINECS: 209-406-4 Merck Index: 12,03460 |
Specification Test Results
Appearance | White Waxy solid |
Infrared spectrum | Conforms |
Assay | ≥95 % |
Water | ≤2 % |
Color of alcohol solution | ≤100 APHA (50 % solids in 1:1 alcohol/water) |
Insoluble matter | ≤300 ppm (50 % in toluene) |
Iodine value | ≤0.25 (on solids basis) |
Acid number | ≤2.5 (on solids basis) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...