2-(Dimethylamino)-ethanol 99%

Supplier: Thermo Scientific Chemicals
Danger

11618-0250 11618-0010 11618-0051 11618-2500
TS11618-0250EA 801 USD
TS11618-0250 TS11618-0010 TS11618-0051 TS11618-2500
2-(Dimethylamino)-ethanol 99%
2-Dimethylaminoethanol
Formula: C₄H₁₁NO
Boiling Pt: 139 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.888 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00002846
CAS Number: 108-01-0
EINECS: 203-542-8
UN: 2051
ADR: 8,II
Merck Index: 13,02863

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4300 (20°C, 589 nm)
Water ≤0.1 %
GC ≥98.5 %
Specific gravity (20°C) 0.885 to 0.889
Isopropyl alcohol ≤0.05 %
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0006 meq/g
Aldehyde (HCHO) ≤0.002 %
Methanol ≤0.05 %
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR