Dimethyl-3-oxoglutarate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate
11570-0250
11570-1000
11570-5000
TS11570-0250EA
43
USD
TS11570-0250
TS11570-1000
TS11570-5000
Dimethyl-3-oxoglutarate 97%
Dimethyl 3-oxoglutarate
Formula:
C₇H₁₀O₅ Boiling Pt: 237…241 °C (760 mmHg) Melting Pt: 16…17 °C Density: 1.18 g/cm³ Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008462 CAS Number: 1830-54-2 EINECS: 217-385-8 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4430 to 1.4450 (20 °C, 589 nm) |
GC | ≥96.0 % |
Visual transition interval | pH 9.0 (blue) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...