Acetonedimethyl acetal 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane

11563-0010 11563-0025 11563-0100 11563-5000
TS11563-0010EA 123 USD
TS11563-0010 TS11563-0025 TS11563-0100 TS11563-5000
Acetonedimethyl acetal 98+%
Acetone-dimethyl acetal
Formula: C₅H₁₂O₂
Boiling Pt: 83 °C (760 mmHg)
Melting Pt: –47 °C
Density: 0.84 g/cm³
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008479
CAS Number: 77-76-9
EINECS: 201-056-0
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3790 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR