Acetonedimethyl acetal 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane
11563-0010
11563-0025
11563-0100
11563-5000
TS11563-0010EA
123
USD
TS11563-0010
TS11563-0025
TS11563-0100
TS11563-5000
Acetonedimethyl acetal 98+%
Acetone-dimethyl acetal
Formula:
C₅H₁₂O₂ Boiling Pt: 83 °C (760 mmHg) Melting Pt: –47 °C Density: 0.84 g/cm³ Flash Pt: –5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008479 CAS Number: 77-76-9 EINECS: 201-056-0 UN: 3271 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3770 to 1.3790 (20°C, 589 nm) |
Water | ≤0.5 % (K.F.) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...